missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-[3-(1H-1,2,4-triazol-1-ylmethyl)benzyl]amine, 97%, Maybridge

$188.76 - $188.76

Chemical Identifiers

CAS 876316-32-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H14N4
Formula Weight 202.26
MDL Number MFCD08435872
Chemical Name or Material N-methyl-N-[3-(1H-1, 2, 4-triazol-1-ylmethyl)benzyl]amine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC45446CB SDS Fisher Scientific Company
CC45446CB
Amber Glass Bottle 250mg Each for $188.76
Specifications Description & Specifications

Chemical Identifiers

876316-32-4
100.0
C11H14N4
MFCD08435872
97%
96.5
97%
202.26
N-methyl-N-[3-(1H-1, 2, 4-triazol-1-ylmethyl)benzyl]amine

Specifications

250mg
96.5
Amber Glass Bottle
C11H14N4
MFCD08435872
97%
876316-32-4
100.0
97%
202.26
N-methyl-N-[3-(1H-1, 2, 4-triazol-1-ylmethyl)benzyl]amine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.