Please login to your online account to display your discounted pricing

N-Methyl-N-(2-thien-2-ylbenzyl)amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material N-methyl-N-(2-thien-2-ylbenzyl)amine
CAS 852180-66-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H13NS
Formula Weight 203.31
MDL Number MFCD07368538
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC35546DE

 
fisher scientific company
CC35546DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC35546CB

 
fisher scientific company
CC35546CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC35546DA

 
fisher scientific company
CC35546DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.31
Quantity 5g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD07368538
Chemical Name or Material N-methyl-N-(2-thien-2-ylbenzyl)amine
Molecular Formula C12H13NS
CAS 852180-66-6
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.