Please login to your online account to display your discounted pricing

N-methyl-N-(2-phenoxybenzyl)amine, Maybridge


Quantity 5g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC49946DE

 View more versions of this product

Catalog No. CC49946DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 213.28
Quantity 5g
Assay 97%
Chemical Name or Material N-Methyl-N-(2-phenoxybenzyl)amine
Assay Percent Range 97%
Molecular Formula C14H15NO
CAS 361394-74-3
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber glass bottle

Specifications

Chemical Name or Material N-Methyl-N-(2-phenoxybenzyl)amine
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15NO
Formula Weight 213.28
Accessories

GHS Signal Word: Danger

SDS & Safety Documents