missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-methyl-N-(2-phenoxybenzyl)amine, Thermo Scientific™
Chemical Identifiers
CAS | 361394-74-3 |
---|---|
Molecular Formula | C14H15NO |
Molecular Weight (g/mol) | 213.28 |
InChI Key | IMIUMEQDSKHKST-UHFFFAOYSA-N |
Synonym | n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride |
PubChem CID | 23512750 |
IUPAC Name | N-methyl-1-(2-phenoxyphenyl)methanamine |
SMILES | CNCC1=CC=CC=C1OC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC49946CB
|
Thermo Scientific Chemicals
CC49946CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC49946DA
|
Thermo Scientific Chemicals
CC49946DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC49946DE
|
Thermo Scientific Chemicals
CC49946DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
361394-74-3 | |
213.28 | |
n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride | |
N-methyl-1-(2-phenoxyphenyl)methanamine |
C14H15NO | |
IMIUMEQDSKHKST-UHFFFAOYSA-N | |
23512750 | |
CNCC1=CC=CC=C1OC2=CC=CC=C2 |
Specifications
361394-74-3 | |
C14H15NO | |
n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride | |
CNCC1=CC=CC=C1OC2=CC=CC=C2 | |
213.28 | |
213.28 | |
N-Methyl-N-(2-phenoxybenzyl)amine |
95% | |
250mg | |
IMIUMEQDSKHKST-UHFFFAOYSA-N | |
N-methyl-1-(2-phenoxyphenyl)methanamine | |
23512750 | |
95% |
Safety and Handling
GHS Signal Word: Danger