Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine, 97%

$149.18 - $483.68

Chemical Identifiers

Chemical Name or Material N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
CAS 120940-43-4
Molecular Formula C8H8F3N3O2
Formula Weight 235.16
MDL Number MFCD00052691
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2482301

  SDS
alfa aesar™
B2482301
0.25g Each for $149.18

AAB2482303

  SDS
alfa aesar™
B2482303
1g Each for $483.68
Description & Specifications

Specifications

Formula Weight 235.16
Chemical Name or Material N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
Melting Point 64° to 67°C
Assay 97%
Quantity 0.25g
Molecular Formula C8H8F3N3O2
MDL Number MFCD00052691
CAS 120940-43-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.