Please login to your online account to display your discounted pricing

N-Methyl-N-[(1-methyl-1H-indazol-3-yl)methyl]amine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC06346CB

 View more versions of this product

Catalog No. CC06346CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 175.23
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material N-methyl-N-[(1-methyl-1H-indazol-3-yl)methyl]amine
Molecular Formula C10H13N3
CAS 124491-38-9
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS Min % 96.5
Packaging Amber Glass Bottle
MDL Number MFCD08060498

Specifications

Chemical Name or Material N-methyl-N-[(1-methyl-1H-indazol-3-yl)methyl]amine
CAS 124491-38-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H13N3
Formula Weight 175.23
MDL Number MFCD08060498
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents