Please login to your online account to display your discounted pricing

N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC03846DA

 View more versions of this product

Catalog No. CC03846DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 175.23
Quantity 1g
Assay 97%
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H13N3
MDL Number MFCD08060627
CAS 137898-62-5
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

Specifications

Chemical Name or Material N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine
CAS 137898-62-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H13N3
Formula Weight 175.23
MDL Number MFCD08060627
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.