missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-L-valine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6525403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
103°C to 104°C | |
3339-44-4 | |
MFCD00238325 | |
n-me-val-ome hcl, n-methyl-l-valine methyl ester hydrochloride, n-me-val-ome . hcl, h-nmeval-ome hydrochloride, methyl n-methyl-l-valinate hydrochloride, n-methyl-l-valine methyl ester hydrochlorid, l-valine, n-methyl-, methyl ester, hydrochloride, methyl 2s-3-methyl-2-methylamino butanoate hydrochloride | |
Cl.CN[C@@H](C(C)C)C(=O)OC | |
181.66 | |
181.66 | |
1 g |
N-Methyl-L-valine methyl ester hydrochloride | |
C7H16ClNO2 | |
3562296 | |
DLLHGHUMLSLOID-RGMNGODLSA-N | |
methyl (2S)-3-methyl-2-(methylamino)butanoate hydrochloride | |
13819431 | |
95% | |
Solid |
Chemical Identifiers
3339-44-4 | |
181.66 | |
DLLHGHUMLSLOID-RGMNGODLSA-N | |
13819431 | |
Cl.CN[C@@H](C(C)C)C(=O)OC |
C7H16ClNO2 | |
MFCD00238325 | |
n-me-val-ome hcl, n-methyl-l-valine methyl ester hydrochloride, n-me-val-ome . hcl, h-nmeval-ome hydrochloride, methyl n-methyl-l-valinate hydrochloride, n-methyl-l-valine methyl ester hydrochlorid, l-valine, n-methyl-, methyl ester, hydrochloride, methyl 2s-3-methyl-2-methylamino butanoate hydrochloride | |
methyl (2S)-3-methyl-2-(methylamino)butanoate hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only