Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-L-valine benzyl ester p-toluenesulfonate, 95%

Chemical Identifiers

Chemical Name or Material N-Methyl-L-valine benzyl ester p-toluenesulfonate
CAS 42492-62-6
Formula Weight 393.5
MDL Number MFCD00238324
Synonym N-Me-Val-Obzl.Tos-OH; N-Methyl-L-valine benzyl ester tosylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6502203

 
alfa aesar™
H6502203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65022MD

 
alfa aesar™
H65022MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 393.5
Chemical Name or Material N-Methyl-L-valine benzyl ester p-toluenesulfonate
Assay 95%
MDL Number MFCD00238324
Synonym N-Me-Val-Obzl.Tos-OH; N-Methyl-L-valine benzyl ester tosylate
Physical Form Powder
CAS 42492-62-6
Storage Note 1 2° to 8°C
Color White
Quantity 1g
Safety and Handling

2° to 8°C