Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-L-proline, 98%

Chemical Identifiers

Chemical Name or Material N-Methyl-L-proline
CAS 475-11-6
Formula Weight 129.16
MDL Number MFCD00011565
Synonym H-N-Me-Pro-OH; (S)-1-Methylpyrrolidine-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6324103

  SDS
alfa aesar™
H6324103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63241MD

  SDS
alfa aesar™
H63241MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.16
Color White to off-white
Chemical Name or Material N-Methyl-L-proline
MDL Number MFCD00011565
Refractive Index 1.5000
CAS 475-11-6
Storage Note 1 -30° to -10°C
Physical Form Powder
Synonym H-N-Me-Pro-OH; (S)-1-Methylpyrrolidine-2-carboxylic acid
Quantity 1g
Assay 98%

Applications
N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.

Solubility
Soluble in water and methanol(both 50 mg/ml-clear-colorless solution)

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

-30° to -10°C