missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-L-proline, 98%

Manufacturer:  Alfa Aesar™ H63241MD

 View more versions of this product

Catalog No. AAH63241MD



Description

Description

Applications
N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.

Solubility
Soluble in water and methanol(both 50 mg/ml-clear-colorless solution)

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

N-Methyl-L-proline
C6H11NO2
n-methyl-l-proline, h-n-me-pro-oh, 1-methylproline, 1-methyl-l-proline, 2s-1-methylpyrrolidine-2-carboxylic acid, s-1-methylpyrrolidine-2-carboxylic acid, l-proline, 1-methyl, n-methyl proline, proline, 1-methyl, s-1-methyl-pyrrolidine-2-carboxylic acid
CWLQUGTUXBXTLF-YFKPBYRVSA-N
(2S)-1-methylpyrrolidine-2-carboxylic acid
643474
Powder
White
475-11-6
MFCD00011565
1.5000
CN1CCCC1C(=O)O
129.159
129.16
98%
250mg

Chemical Identifiers

475-11-6
129.159
CWLQUGTUXBXTLF-YFKPBYRVSA-N
643474
CN1CCCC1C(=O)O
C6H11NO2
MFCD00011565
n-methyl-l-proline, h-n-me-pro-oh, 1-methylproline, 1-methyl-l-proline, 2s-1-methylpyrrolidine-2-carboxylic acid, s-1-methylpyrrolidine-2-carboxylic acid, l-proline, 1-methyl, n-methyl proline, proline, 1-methyl, s-1-methyl-pyrrolidine-2-carboxylic acid
(2S)-1-methylpyrrolidine-2-carboxylic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents