missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-L-isoleucine hydrochloride, 95%

$58.15 - $862.04

Chemical Identifiers

CAS 66866-68-0
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD06801364
InChI Key MMBGZHHBWVCZFK-GEMLJDPKSA-N
Synonym 2s,3s-3-methyl-2-methylamino pentanoic acid hydrochloride, n-methyl-l-isoleucine hydrochloride, n-me-ile-oh, methyl l-+-isoleucine hydrochloride, n-alpha-methyl-l-isoleucine hydrochloride, n-methyl-l-isoleucine-hydrogen chloride 1/1
PubChem CID 56777139
IUPAC Name (2S,3S)-3-methyl-2-(methylamino)pentanoic acid;hydrochloride
SMILES CCC(C)C(C(=O)O)NC.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6597903 View Documents Alfa Aesar™
H6597903
1g Each for $58.15
 
AAH6597906 View Documents Alfa Aesar™
H6597906
5g Each for $215.89
 
AAH6597914 View Documents Alfa Aesar™
H6597914
25g Each for $862.04
 
Specifications

Chemical Identifiers

66866-68-0
181.66
MMBGZHHBWVCZFK-GEMLJDPKSA-N
56777139
CCC(C)C(C(=O)O)NC.Cl
C7H16ClNO2
MFCD06801364
2s,3s-3-methyl-2-methylamino pentanoic acid hydrochloride, n-methyl-l-isoleucine hydrochloride, n-me-ile-oh, methyl l-+-isoleucine hydrochloride, n-alpha-methyl-l-isoleucine hydrochloride, n-methyl-l-isoleucine-hydrogen chloride 1/1
(2S,3S)-3-methyl-2-(methylamino)pentanoic acid;hydrochloride

Specifications

66866-68-0
MFCD06801364
2s,3s-3-methyl-2-methylamino pentanoic acid hydrochloride, n-methyl-l-isoleucine hydrochloride, n-me-ile-oh, methyl l-+-isoleucine hydrochloride, n-alpha-methyl-l-isoleucine hydrochloride, n-methyl-l-isoleucine-hydrogen chloride 1/1
CCC(C)C(C(=O)O)NC.Cl
181.66
181.66
95%
1g
C7H16ClNO2
3689642
MMBGZHHBWVCZFK-GEMLJDPKSA-N
(2S,3S)-3-methyl-2-(methylamino)pentanoic acid;hydrochloride
56777139
Solid
White
N-Methyl-L-isoleucine hydrochloride