missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-L-arginine

$407.12 - $1,568.34

Chemical Identifiers

CAS 2480-28-6
Molecular Formula C7H16N4O2
Molecular Weight (g/mol) 188.231
MDL Number MFCD12828775
InChI Key NTWVQPHTOUKMDI-YFKPBYRVSA-N
Synonym n-methyl-l-arginine, n2-methyl-l-arginine, arginine, n2-methyl-, l, unii-88rbj4ze9r, 88rbj4ze9r, l-monomethyl-l-arginine, s-5-guanidino-2-methylamino pentanoic acid, n 2-methyl-l-arginine, n~5~-diaminomethylene-n~2~-methylornithine, n-mearg
PubChem CID 73046
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-(methylamino)pentanoic acid
SMILES CNC(CCCN=C(N)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62431MA View Documents Alfa Aesar™
J62431MA
10mg Each for $407.12
 
AAJ62431MF View Documents Alfa Aesar™
J62431MF
50mg Each for $1,568.34
 
Description

Description

Applications
N-Methyl-L-arginine is used as a selective inhibitor of L-arginine uptake. It comes under the category of ion pump transporters.

Solubility
Soluble to 100 mM in water

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. For obtaining a higher solubility , please warm the tube at 37°C and shake it in the ultrasonic bath for a while. Stock solution can be stored below -20°C for several months.
Specifications

Chemical Identifiers

2480-28-6
188.231
NTWVQPHTOUKMDI-YFKPBYRVSA-N
73046
CNC(CCCN=C(N)N)C(=O)O
C7H16N4O2
MFCD12828775
n-methyl-l-arginine, n2-methyl-l-arginine, arginine, n2-methyl-, l, unii-88rbj4ze9r, 88rbj4ze9r, l-monomethyl-l-arginine, s-5-guanidino-2-methylamino pentanoic acid, n 2-methyl-l-arginine, n~5~-diaminomethylene-n~2~-methylornithine, n-mearg
(2S)-5-(diaminomethylideneamino)-2-(methylamino)pentanoic acid

Specifications

2480-28-6
MFCD12828775
Soluble to 100 mM in water
CNC(CCCN=C(N)N)C(=O)O
188.231
188.23
White
N-Methyl-L-arginine
C7H16N4O2
n-methyl-l-arginine, n2-methyl-l-arginine, arginine, n2-methyl-, l, unii-88rbj4ze9r, 88rbj4ze9r, l-monomethyl-l-arginine, s-5-guanidino-2-methylamino pentanoic acid, n 2-methyl-l-arginine, n~5~-diaminomethylene-n~2~-methylornithine, n-mearg
NTWVQPHTOUKMDI-YFKPBYRVSA-N
(2S)-5-(diaminomethylideneamino)-2-(methylamino)pentanoic acid
73046
Powder
10mg