missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-L-alanine, 98%

$28.07 - $89.30

Chemical Identifiers

CAS 3913-67-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00037241
InChI Key GDFAOVXKHJXLEI-VKHMYHEASA-N
Synonym n-methyl-l-alanine, alanine, n-methyl-, l, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine
PubChem CID 5288725
ChEBI CHEBI:17519
IUPAC Name (2S)-2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60103MD View Documents Alfa Aesar™
H60103MD
250mg Each for $28.07
 
AAH6010303 View Documents Alfa Aesar™
H6010303
1g Each for $89.30
 
Specifications

Chemical Identifiers

3913-67-5
103.121
GDFAOVXKHJXLEI-VKHMYHEASA-N
5288725
(2S)-2-(methylamino)propanoic acid
C4H9NO2
MFCD00037241
n-methyl-l-alanine, alanine, n-methyl-, l, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine
CHEBI:17519
CC(C(=O)O)NC

Specifications

3913-67-5
MFCD00037241
n-methyl-l-alanine, alanine, n-methyl-, l, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine
CC(C(=O)O)NC
103.121
CHEBI:17519
98%
N-Methyl-L-alanine
C4H9NO2
1720922
GDFAOVXKHJXLEI-VKHMYHEASA-N
(2S)-2-(methylamino)propanoic acid
5288725
103.12
250mg