missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-DL-alanine, 98%

Manufacturer:  Alfa Aesar™ H6528736

 View more versions of this product

Catalog No. AAH6528736



Specifications

Specifications

N-Methyl-DL-alanine
C4H9NO2
2-methylamino propanoic acid, a-methyl-dl-alanine, alanine, n-methyl-, l, dl-n-methylalanine, h-n-me-dl-ala-oh, me-dl-ala-oh, n-, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, n-methyl cdl-alanine, n-methyl-dl-alanine
CC(C(=O)O)NC
103.121
103.12
98%
500g
600-21-5
MFCD00063136
GDFAOVXKHJXLEI-UHFFFAOYSA-N
2-(methylamino)propanoic acid
4377
Crystalline Powder
White

Chemical Identifiers

600-21-5
103.121
GDFAOVXKHJXLEI-UHFFFAOYSA-N
4377
CC(C(=O)O)NC
C4H9NO2
MFCD00063136
2-methylamino propanoic acid, a-methyl-dl-alanine, alanine, n-methyl-, l, dl-n-methylalanine, h-n-me-dl-ala-oh, me-dl-ala-oh, n-, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, n-methyl cdl-alanine, n-methyl-dl-alanine
2-(methylamino)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents