missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-D-phenylalanine, 97%

$47.04 - $462.64

Chemical Identifiers

CAS 56564-52-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00069560
InChI Key SCIFESDRCALIIM-SECBINFHSA-N
Synonym r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
PubChem CID 6427037
IUPAC Name (2R)-2-(methylamino)-3-phenylpropanoic acid
SMILES CNC(CC1=CC=CC=C1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65675MD View Documents Alfa Aesar™
H65675MD
250mg Each for $47.04
 
AAH6567503 View Documents Alfa Aesar™
H6567503
1g Each for $124.14
 
AAH6567506 View Documents Alfa Aesar™
H6567506
5g Each for $462.64
 
Specifications

Chemical Identifiers

56564-52-4
179.219
SCIFESDRCALIIM-SECBINFHSA-N
6427037
CNC(CC1=CC=CC=C1)C(=O)O
C10H13NO2
MFCD00069560
r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
(2R)-2-(methylamino)-3-phenylpropanoic acid

Specifications

56564-52-4
MFCD00069560
r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
CNC(CC1=CC=CC=C1)C(=O)O
179.219
179.22
97%
N-Methyl-D-phenylalanine
C10H13NO2
3199933
SCIFESDRCALIIM-SECBINFHSA-N
(2R)-2-(methylamino)-3-phenylpropanoic acid
6427037
Solid
250mg