Please login to your online account to display your discounted pricing

N-Methyl-D-aspartic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC329190010

 View more versions of this product

Catalog No. AC329190010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5% (total base)
Formula Weight 147.1
Quantity 1g
Color White to off-white
RTECS Number CI9457000
Specific Rotation Condition − 28.00 (20.00°C c=1,HCl 6N)
Merck Index 15, 6749
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 LIGHT SENSITIVE
CAS Min % 97.5
Specific Rotation −26° to −30° (20°C, 589nm) (c=1, 6 N HCl)
Synonym NMDA
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H9NO4
Name Note 98%
MDL Number MFCD00004226
CAS 6384-92-5
Chemical Name or Material N-Methyl-D-aspartic acid

Specifications

Chemical Name or Material N-Methyl-D-aspartic acid
Name Note 98%
CAS 6384-92-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (total base)
Assay Percent Range 98%
Molecular Formula C5H9NO4
Formula Weight 147.1
MDL Number MFCD00004226
Synonym NMDA
Accessories

LIGHT SENSITIVE

SDS & Safety Documents