Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-D-alanine, 98%

$135.61 - $543.36

Chemical Identifiers

Chemical Name or Material N-Methyl-D-alanine
CAS 29475-64-7
Formula Weight 103.12
MDL Number MFCD00063135
Beilstein 1720923
Synonym H-N-Me-D-Ala-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6584006

 
alfa aesar™
H6584006
5g Each for $135.61

AAH6584014

 
alfa aesar™
H6584014
25g Each for $543.36
Description & Specifications

Specifications

Formula Weight 103.12
Chemical Name or Material N-Methyl-D-alanine
CAS 29475-64-7
Assay 98%
MDL Number MFCD00063135
Synonym H-N-Me-D-Ala-OH
Physical Form Powder
Beilstein 1720923
Color White
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C