Learn More
n-methyl-(5-pyrid-3-ylthien-2-yl)methylamine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 837376-49-5 |
---|---|
Molecular Formula | C11H12N2S |
Molecular Weight (g/mol) | 204.291 |
MDL Number | MFCD11109328 |
InChI Key | ZSTXLHXHAPANSL-UHFFFAOYSA-N |
Synonym | 2-methylamino methyl-5-pyridin-3-ylthiophene, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, methyl 5-3-pyridyl 2-thienyl methyl amine, methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, nicotine 3-heteroaromatic analogue 1b |
PubChem CID | 11321674 |
IUPAC Name | N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine |
SMILES | CNCC1=CC=C(S1)C2=CN=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC66846CB
|
Fisher Scientific Company
CC66846CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC66846DA
|
Fisher Scientific Company
CC66846DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
837376-49-5 | |
204.291 | |
ZSTXLHXHAPANSL-UHFFFAOYSA-N | |
11321674 | |
CNCC1=CC=C(S1)C2=CN=CC=C2 |
C11H12N2S | |
MFCD11109328 | |
2-methylamino methyl-5-pyridin-3-ylthiophene, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, methyl 5-3-pyridyl 2-thienyl methyl amine, methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, nicotine 3-heteroaromatic analogue 1b | |
N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine |
Specifications
837376-49-5 | |
100.0 | |
C11H12N2S | |
Amber Glass Bottle | |
250mg | |
CNCC1=CC=C(S1)C2=CN=CC=C2 | |
204.291 | |
204.3 | |
N-methyl-(5-pyrid-3-ylthien-2-yl)methylamine |
96.5 | |
97% | |
MFCD11109328 | |
2-methylamino methyl-5-pyridin-3-ylthiophene, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, methyl 5-3-pyridyl 2-thienyl methyl amine, methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, nicotine 3-heteroaromatic analogue 1b | |
ZSTXLHXHAPANSL-UHFFFAOYSA-N | |
N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine | |
11321674 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger