Please login to your online account to display your discounted pricing

n-methyl-[(5-piperidinothien-2-yl)methyl]amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC72646DA

 View more versions of this product

Catalog No. CC72646DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 210.34
Quantity 1g
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
CAS 953409-06-8
CAS Min % 96.5
Chemical Name or Material N-methyl-[(5-piperidinothien-2-yl)methyl]amine
Assay Percent Range 97%
Molecular Formula C11H18N2S
MDL Number MFCD11506355

Specifications

Chemical Name or Material N-methyl-[(5-piperidinothien-2-yl)methyl]amine
CAS 953409-06-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H18N2S
Formula Weight 210.34
MDL Number MFCD11506355