Please login to your online account to display your discounted pricing

n-methyl-[(5-piperidinothien-2-yl)methyl]amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material N-methyl-[(5-piperidinothien-2-yl)methyl]amine
CAS 953409-06-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H18N2S
Formula Weight 210.34
MDL Number MFCD11506355
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC72646DA

  SDS
fisher scientific company
CC72646DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC72646CB

  SDS
fisher scientific company
CC72646CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.34
Quantity 1g
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
CAS Min % 96.5
CAS 953409-06-8
MDL Number MFCD11506355
Assay Percent Range 97%
Chemical Name or Material N-methyl-[(5-piperidinothien-2-yl)methyl]amine
Molecular Formula C11H18N2S