missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-4-nitrophthalimide, TCI America™

$47.36 - $146.67

Chemical Identifiers

CAS 41663-84-7
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD00454263
InChI Key JBCHWGTZAAZJKG-UHFFFAOYSA-N
PubChem CID 92321
IUPAC Name 2-methyl-5-nitroisoindole-1,3-dione
SMILES CN1C(=O)C2=C(C1=O)C=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M131025G View Documents TCI America
M131025G
25g Each for $146.67
 
M13105G View Documents TCI America
M13105G
5g Each for $47.36
 
Specifications

Chemical Identifiers

41663-84-7
206.157
JBCHWGTZAAZJKG-UHFFFAOYSA-N
2-methyl-5-nitroisoindole-1,3-dione
C9H6N2O4
MFCD00454263
92321
CN1C(=O)C2=C(C1=O)C=C(C=C2)[N+](=O)[O-]

Specifications

Yellow
25g
C9H6N2O4
JBCHWGTZAAZJKG-UHFFFAOYSA-N
2-methyl-5-nitroisoindole-1,3-dione
92321
Crystal-Powder at 20°C
N-Methyl-4-nitrophthalimide
178°C
41663-84-7
MFCD00454263
CN1C(=O)C2=C(C1=O)C=C(C=C2)[N+](=O)[O-]
206.157
206.16
≥99.0% (HPLC)
Safety and Handling

Safety and Handling

RTECSNumber : NR3398541

TSCA : Yes