Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-4-nitroaniline, 97%

Chemical Identifiers

Chemical Name or Material N-Methyl-4-nitroaniline
CAS 100-15-2
Molecular Formula C7H8N2O2
Formula Weight 152.15g/mol
MDL Number MFCD00007305
UN Number UN2811
Beilstein 637920
Synonym 4-Methylaminonitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1554814

 
alfa aesar™
A15548-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1554822

 
alfa aesar™
A15548-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 147° to 154°C
Quantity 25g
Odor Characteristic
Chemical Name or Material N-Methyl-4-nitroaniline
CAS 100-15-2
Assay 97%
Molecular Formula C7H8N2O2
Formula Weight 152.15g/mol
MDL Number MFCD00007305
UN Number UN2811
Beilstein 637920
Synonym 4-Methylaminonitrobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA