Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-4-nitro-alpha-toluenesulfonamide, 98%

$75.11 - $221.66

Chemical Identifiers

Chemical Name or Material N-Methyl-4-nitro-alpha-toluenesulfonamide
CAS 85952-29-0
Molecular Formula C8H10N2O4S
Formula Weight 230.24
MDL Number MFCD00277775
Synonym N-Methyl-4-nitrobenzylsulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2579503

 
alfa aesar™
H2579503
1g Each for $75.11

AAH2579506

 
alfa aesar™
H2579506
5g Each for $221.66
Description & Specifications

Specifications

Formula Weight 230.24
Chemical Name or Material N-Methyl-4-nitro-alpha-toluenesulfonamide
Melting Point 151° to 153°C
Assay 98%
Quantity 1g
Molecular Formula C8H10N2O4S
MDL Number MFCD00277775
CAS 85952-29-0
Synonym N-Methyl-4-nitrobenzylsulfonamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.