Please login to your online account to display your discounted pricing

N-Methyl(4-methylthien-2-yl)methylamine 97+%, Maybridge

Chemical Identifiers

Chemical Name or Material N-Methyl(4-methylthien-2-yl)methylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H11NS
Formula Weight 141.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC30646CB

  SDS
fisher scientific company
CC30646CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC30646DA

  SDS
fisher scientific company
CC30646DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.24
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
CAS 886851-27-0
Assay Percent Range 97%
Chemical Name or Material N-Methyl(4-methylthien-2-yl)methylamine
Molecular Formula C7H11NS