Learn More
N-Methyl-(4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1031843-28-3 |
---|---|
Molecular Formula | C12H14N2O |
Molecular Weight (g/mol) | 202.257 |
MDL Number | MFCD11109322 |
InChI Key | XCBYKJIAOCVNGY-UHFFFAOYSA-N |
Synonym | n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine |
PubChem CID | 33589464 |
IUPAC Name | N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine |
SMILES | CC1=C(OC(=N1)C2=CC=CC=C2)CNC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC18146CB
|
Thermo Scientific Chemicals
CC18146CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC18146DA
|
Thermo Scientific Chemicals
CC18146DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1031843-28-3 | |
202.257 | |
XCBYKJIAOCVNGY-UHFFFAOYSA-N | |
33589464 | |
CC1=C(OC(=N1)C2=CC=CC=C2)CNC |
C12H14N2O | |
MFCD11109322 | |
n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine | |
N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine |
Specifications
1031843-28-3 | |
100.0 | |
Amber Glass Bottle | |
MFCD11109322 | |
n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine | |
CC1=C(OC(=N1)C2=CC=CC=C2)CNC | |
202.257 | |
202.26 | |
N-Methyl-(4-methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine |
96.5 | |
97% | |
C12H14N2O | |
250mg | |
XCBYKJIAOCVNGY-UHFFFAOYSA-N | |
N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine | |
33589464 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger