Please login to your online account to display your discounted pricing

N-Methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride, Tech., Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC54296DA

 View more versions of this product

Catalog No. CC54296DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 242.57
Quantity 1g
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Chemical Name or Material N-methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride
Molecular Formula C6H8BrNS·ClH
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD09817483
CAS 944450-82-2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material N-methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride
CAS 944450-82-2
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C6H8BrNS·ClH
Formula Weight 242.57
MDL Number MFCD09817483
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents