Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-4-(2-chloroacetamido)benzoic acid, 98%

$225.36 - $225.36

Chemical Identifiers

Chemical Name or Material N-Methyl-4-(2-chloroacetamido)benzoic acid
CAS 147149-44-8
Molecular Formula C10H10ClNO3
Formula Weight 227.65g/mol
MDL Number MFCD02093984
Synonym 4'-Carboxy-N-methyl-2-chloroacetanilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1802706

 
alfa aesar™
L18027-06
5g Each for $225.36
Description & Specifications

Specifications

Melting Point 188° to 190°C
Quantity 5g
Chemical Name or Material N-Methyl-4-(2-chloroacetamido)benzoic acid
CAS 147149-44-8
Assay 98%
Molecular Formula C10H10ClNO3
Formula Weight 227.65g/mol
MDL Number MFCD02093984
Synonym 4'-Carboxy-N-methyl-2-chloroacetanilide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.