missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-3-(trifluoromethyl)benzylamine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6388806

 View more versions of this product

Catalog No. AAH6388806



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Methyl-3-(trifluoromethyl)benzylamine
Colorless
MFCD04115410
UN2735
JFLPPELZYKHKQZ-UHFFFAOYSA-N
N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
485414
95%
90390-07-1
C9H10F3N
5 g
n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
CNCC1=CC(=CC=C1)C(F)(F)F
189.181
189.18
Oil

Chemical Identifiers

90390-07-1
189.181
JFLPPELZYKHKQZ-UHFFFAOYSA-N
485414
CNCC1=CC(=CC=C1)C(F)(F)F
C9H10F3N
MFCD04115410
n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only