missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-3-pyrimidin-2-ylbenzylamine, 97%, Maybridge

$97.04 - $97.04

Chemical Identifiers

CAS 886851-49-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H13N3
Formula Weight 199.26
MDL Number MFCD09702386
Chemical Name or Material N-methyl-3-pyrimidin-2-ylbenzylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC56346DA SDS Fisher Scientific Company
CC56346DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC56346CB SDS Fisher Scientific Company
CC56346CB
Amber Glass Bottle 250mg Each for $97.04
Specifications Description & Specifications

Chemical Identifiers

886851-49-6
100.0
C12H13N3
MFCD09702386
97%
96.5
97%
199.26
N-methyl-3-pyrimidin-2-ylbenzylamine

Specifications

199.26
100.0
96.5
Amber Glass Bottle
97%
97%
1g
C12H13N3
MFCD09702386
886851-49-6
N-methyl-3-pyrimidin-2-ylbenzylamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger