Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-3-pyridinemethylamine, 98%

$76.19 - $317.09

Chemical Identifiers

Chemical Name or Material N-Methyl-3-pyridinemethylamine
CAS 20173-04-0
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00023610
Synonym alpha-Methylamino-3-picoline; N-Methyl-3-pyridylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5026203

 
alfa aesar™
H5026203
1g Each for $76.19

AAH5026206

 
alfa aesar™
H5026206
5g Each for $317.09
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material N-Methyl-3-pyridinemethylamine
Boiling Point 93° to 95°C (9mmHg)
Assay 98%
Quantity 1g
Molecular Formula C7H10N2
MDL Number MFCD00023610
CAS 20173-04-0
Synonym alpha-Methylamino-3-picoline; N-Methyl-3-pyridylmethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.