missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-3-nitroaniline, 97%

Manufacturer:  Alfa Aesar™ H5519603

 View more versions of this product

Catalog No. AAH5519603



Specifications

Specifications

N-Methyl-3-nitroaniline
64°C to 68°C
619-26-1
MFCD00963641
IKSRCCUOUJJGAU-UHFFFAOYSA-N
N-methyl-3-nitroaniline
219622
Powder
White to Brown
1g
C7H8N2O2
3-nitro-n-methylaniline
CNC1=CC(=CC=C1)[N+](=O)[O-]
152.153
152.15
97%

Chemical Identifiers

619-26-1
152.153
IKSRCCUOUJJGAU-UHFFFAOYSA-N
219622
CNC1=CC(=CC=C1)[N+](=O)[O-]
C7H8N2O2
MFCD00963641
3-nitro-n-methylaniline
N-methyl-3-nitroaniline
Safety and Handling

Safety and Handling

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

TSCA : No

Recommended Storage : Ambient temperatures

Documents