missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-3-methylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H5629903

 View more versions of this product

Catalog No. AAH5629903



Specifications

Specifications

N-Methyl-3-methylbenzamide
74786-81-5
MFCD01211937
WKSODUAHKWBHQZ-UHFFFAOYSA-N
N,3-dimethylbenzamide
966002
97%
1g
C9H11NO
n-methyl-3-methylbenzamide, 3,n-dimethylbenzamide, benzamide,n,3-dimethyl, benzamide, n,3-dimethyl-9ci, n-methyl 3-methylphenyl carboxamide
CC1=CC=CC(=C1)C(=O)NC
149.193
149.18

Chemical Identifiers

74786-81-5
149.193
WKSODUAHKWBHQZ-UHFFFAOYSA-N
966002
CC1=CC=CC(=C1)C(=O)NC
C9H11NO
MFCD01211937
n-methyl-3-methylbenzamide, 3,n-dimethylbenzamide, benzamide,n,3-dimethyl, benzamide, n,3-dimethyl-9ci, n-methyl 3-methylphenyl carboxamide
N,3-dimethylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents