Please login to your online account to display your discounted pricing

N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC62646CB

 View more versions of this product

Catalog No. CC62646CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.28
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine
Assay Percent Range 95%
Molecular Formula C13H15N3O
MDL Number MFCD09817528
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
CAS 912569-66-5

Specifications

Chemical Name or Material N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine
CAS 912569-66-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 95%
Molecular Formula C13H15N3O
Formula Weight 229.28
MDL Number MFCD09817528
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

SDS & Safety Documents