Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-3,4,5-trimethoxybenzylamine, 97%

$152.39 - $152.39

Chemical Identifiers

Chemical Name or Material N-Methyl-3,4,5-trimethoxybenzylamine
CAS 58780-82-8
Molecular Formula C11H17NO3
Formula Weight 211.26
MDL Number MFCD00798597
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2099606

 
alfa aesar™
B2099606
5g Each for $152.39
Description & Specifications

Specifications

Formula Weight 211.26
Chemical Name or Material N-Methyl-3,4,5-trimethoxybenzylamine
Assay 97%
Quantity 5g
Molecular Formula C11H17NO3
MDL Number MFCD00798597
CAS 58780-82-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.