Learn More
N-Methyl-3-(1H-pyrazol-1-ylmethyl)benzylamine, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC56146DA
Specifications
N-methyl-3-(1H-pyrazol-1-ylmethyl)benzylamine | |
89.5 | |
90% | |
C12H15N3 | |
1g | |
RXKBCAGRFVIQTK-UHFFFAOYSA-N | |
N-methyl-1-[3-(pyrazol-1-ylmethyl)phenyl]methanamine | |
18525881 | |
90% |
892502-10-2 | |
100.0 | |
Amber Glass Bottle | |
MFCD08690303 | |
n-methyl-3-1h-pyrazol-1-ylmethyl benzylamine, methyl 3-pyrazol-1-ylmethyl phenyl methyl amine, methyl 3-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-3-1h-pyrazol-1-ylmethyl, 1-3-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-3-1h-pyrazol-1-yl methyl phenyl methanamine | |
CNCC1=CC=CC(=C1)CN2C=CC=N2 | |
201.273 | |
201.27 |
Chemical Identifiers
892502-10-2 | |
201.273 | |
RXKBCAGRFVIQTK-UHFFFAOYSA-N | |
18525881 | |
CNCC1=CC=CC(=C1)CN2C=CC=N2 |
C12H15N3 | |
MFCD08690303 | |
n-methyl-3-1h-pyrazol-1-ylmethyl benzylamine, methyl 3-pyrazol-1-ylmethyl phenyl methyl amine, methyl 3-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-3-1h-pyrazol-1-ylmethyl, 1-3-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-3-1h-pyrazol-1-yl methyl phenyl methanamine | |
N-methyl-1-[3-(pyrazol-1-ylmethyl)phenyl]methanamine |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.