Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-2-(trifluoromethyl)aniline, 96%

$219.99 - $219.99

Chemical Identifiers

Chemical Name or Material N-Methyl-2-(trifluoromethyl)aniline
CAS 14925-10-1
Molecular Formula C8H8F3N
Formula Weight 175.16
MDL Number MFCD00052196
Beilstein 2641847
Synonym 2-(Methylamino)benzotrifluoride; 2-Trifluoromethyl-N-methylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0980106

  SDS
alfa aesar™
L0980106
5g Each for $219.99
Description & Specifications

Specifications

Formula Weight 175.16
Chemical Name or Material N-Methyl-2-(trifluoromethyl)aniline
Assay 96%
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2-(Methylamino)benzotrifluoride; 2-Trifluoromethyl-N-methylaniline
Beilstein 2641847
MDL Number MFCD00052196
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Quantity 5g
Flash Point 64°C (147°F)
Molecular Formula C8H8F3N
Boiling Point 177°C
Health Hazard 1 Warning
CAS 14925-10-1
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.