missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-2-(trifluoromethyl)aniline, 96%

Manufacturer:  Alfa Aesar™ L0980103

 View more versions of this product

Catalog No. AAL0980103



Specifications

Specifications

N-Methyl-2-(trifluoromethyl)aniline
177°C
14925-10-1
MFCD00052196
n-methyl-2-trifluoromethyl aniline, 2-methylamino benzotrifluoride, 2-trifluoromethyl-n-methylaniline, benzenamine, n-methyl-2-trifluoromethyl, n-methyl-n-2-trifluoromethyl phenyl amine, methyl 2-trifluoromethyl phenyl amine, 2-methylamino benzotrifluride, 2-trifluoromethyl-n-methylaniline, benzenamine,n-methyl-2-trifluoromethyl, 2-trifluoromethyl-n-methylaniline
CNC1=CC=CC=C1C(F)(F)F
175.154
175.16
64°C (147°F)
1g
C8H8F3N
2641847
RNRDHZGWBBEZBK-UHFFFAOYSA-N
N-methyl-2-(trifluoromethyl)aniline
2760894
96%

Chemical Identifiers

14925-10-1
175.154
RNRDHZGWBBEZBK-UHFFFAOYSA-N
2760894
CNC1=CC=CC=C1C(F)(F)F
C8H8F3N
MFCD00052196
n-methyl-2-trifluoromethyl aniline, 2-methylamino benzotrifluoride, 2-trifluoromethyl-n-methylaniline, benzenamine, n-methyl-2-trifluoromethyl, n-methyl-n-2-trifluoromethyl phenyl amine, methyl 2-trifluoromethyl phenyl amine, 2-methylamino benzotrifluride, 2-trifluoromethyl-n-methylaniline, benzenamine,n-methyl-2-trifluoromethyl, 2-trifluoromethyl-n-methylaniline
N-methyl-2-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents