missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-2-(phenoxymethyl)benzylamine, 97%, Maybridge

Chemical Identifiers

CAS 892502-17-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C15H17NO
Formula Weight 227.31
MDL Number MFCD09025894
Chemical Name or Material N-methyl-2-(phenoxymethyl)benzylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC58546CB SDS Fisher Scientific Company
CC58546CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58546DA SDS Fisher Scientific Company
CC58546DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

892502-17-9
100.0
C15H17NO
MFCD09025894
97%
96.5
97%
227.31
N-methyl-2-(phenoxymethyl)benzylamine

Specifications

250mg
96.5
97%
227.31
N-methyl-2-(phenoxymethyl)benzylamine
892502-17-9
100.0
C15H17NO
MFCD09025894
97%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.