Please login to your online account to display your discounted pricing

N-methyl-(2-methyl-1,3-oxazol-4-yl)methylamine, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC57946CB

 View more versions of this product

Catalog No. CC57946CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.16
Quantity 250mg
Assay 97%
Molecular Formula C6H10N2O
CAS 1065073-46-2
Packaging Amber Glass Bottle
Chemical Name or Material N-methyl-(2-methyl-1, 3-oxazol-4-yl)methylamine
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material N-methyl-(2-methyl-1, 3-oxazol-4-yl)methylamine
Assay 97%
Molecular Formula C6H10N2O
Formula Weight 126.16
Accessories

GHS Signal Word: Danger

SDS & Safety Documents