Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-2-biphenylmethylamine, 99%

$117.61 - $410.99

Chemical Identifiers

Chemical Name or Material N-Methyl-2-biphenylmethylamine
CAS 13737-31-0
Molecular Formula C14H15N
Formula Weight 197.28
MDL Number MFCD00598943
Synonym 2-Biphenylyl-N-methylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5076503

  SDS
alfa aesar™
H5076503
1g Each for $410.99

AAH50765MD

  SDS
alfa aesar™
H50765MD
250mg Each for $117.61
Description & Specifications

Specifications

Formula Weight 197.28
Chemical Name or Material N-Methyl-2-biphenylmethylamine
Assay 99%
Quantity 1g
Molecular Formula C14H15N
MDL Number MFCD00598943
CAS 13737-31-0
Synonym 2-Biphenylyl-N-methylmethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.