missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-2-(4-methoxyphenyl)ethylamine, TCI America™

$123.75 - $734.88

Chemical Identifiers

CAS 4091-50-3
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00870496
InChI Key JCMWSVNNSPUNER-UHFFFAOYSA-N
Synonym 4-Methoxy-N-methylphenethylamine, 4-[2-(Methylamino)ethyl]anisole
PubChem CID 104735
ChEBI CHEBI:75143
IUPAC Name 2-(4-methoxyphenyl)-N-methylethanamine
SMILES CNCCC1=CC=C(C=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M255120MG View Documents TCI America
M255120MG
20mg Each for $123.75
 
M2551200MG View Documents TCI America
M2551200MG
200mg Each for $734.88
 
Specifications

Chemical Identifiers

4091-50-3
165.236
JCMWSVNNSPUNER-UHFFFAOYSA-N
104735
2-(4-methoxyphenyl)-N-methylethanamine
C10H15NO
MFCD00870496
4-Methoxy-N-methylphenethylamine, 4-[2-(Methylamino)ethyl]anisole
CHEBI:75143
CNCCC1=CC=C(C=C1)OC

Specifications

83°C
20mg
C10H15NO
4-Methoxy-N-methylphenethylamine, 4-[2-(Methylamino)ethyl]anisole
CNCCC1=CC=C(C=C1)OC
165.236
CHEBI:75143
Clear Liquid at 20°C
N-Methyl-2-(4-methoxyphenyl)ethylamine
Yellow
4091-50-3
MFCD00870496
JCMWSVNNSPUNER-UHFFFAOYSA-N
2-(4-methoxyphenyl)-N-methylethanamine
104735
165.24
≥95.0% (GC,T)
Safety and Handling

Safety and Handling

RTECSNumber : SH8110000

TSCA : No