Please login to your online account to display your discounted pricing

N-Methyl-[(2,3-dihydrobenzo[b]furan-7-yl)methyl]amine, 90%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC00946CB

 View more versions of this product

Catalog No. CC00946CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 163.22
Quantity 250mg
Assay 90%
CAS Max % 100.0
CAS 389845-43-6
Packaging Amber Glass Bottle
Chemical Name or Material N-methyl-[(2, 3-dihydrobenzo[b]furan-7-yl)methyl]amine
Assay Percent Range 90%
Molecular Formula C10H13NO
MDL Number MFCD10700046
CAS Min % 89.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-methyl-[(2, 3-dihydrobenzo[b]furan-7-yl)methyl]amine
CAS 389845-43-6
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C10H13NO
Formula Weight 163.22
MDL Number MFCD10700046
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents