missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-2-(2-pyridyl)ethylamine methanesulfonate

$15.09 - $15.09

Chemical Identifiers

CAS 380416-14-8
Molecular Formula C9H16N2O3S
Formula Weight 232.3
MDL Number MFCD00143688
Synonym N-Methyl-2-pyridin-2-ylethylamine methanesulfonate
Chemical Name or Material N-Methyl-2-(2-pyridyl)ethylamine methanesulfonate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5019003 SDS Alfa Aesar™
H5019003
1g Each for $15.09
AAH5019006 SDS Alfa Aesar™
H5019006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

380416-14-8
232.3
N-Methyl-2-pyridin-2-ylethylamine methanesulfonate
C9H16N2O3S
MFCD00143688
N-Methyl-2-(2-pyridyl)ethylamine methanesulfonate

Specifications

380416-14-8
C9H16N2O3S
MFCD00143688
N-Methyl-2-(2-pyridyl)ethylamine methanesulfonate
1g
232.3
N-Methyl-2-pyridin-2-ylethylamine methanesulfonate

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning