Please login to your online account to display your discounted pricing

N-Methyl-2-(1H-1,2,4-triazol-1-ylmethyl)benzylamine, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC47546CB

 View more versions of this product

Catalog No. CC47546CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.26
Quantity 250mg
Assay 97%
CAS Max % 100.0
MDL Number MFCD09025848
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C11H14N4
Chemical Name or Material N-methyl-2-(1H-1, 2, 4-triazol-1-ylmethyl)benzylamine
CAS Min % 96.5
CAS 906352-64-5
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material N-methyl-2-(1H-1, 2, 4-triazol-1-ylmethyl)benzylamine
CAS 906352-64-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H14N4
Formula Weight 202.26
MDL Number MFCD09025848
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents