Please login to your online account to display your discounted pricing

N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC75946DA

 View more versions of this product

Catalog No. CC75946DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.24
Quantity 1g
Assay 97%
CAS 103456607
Chemical Name or Material N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine
Molecular Formula C11H13N3
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine
Assay 97%
Molecular Formula C11H13N3
Formula Weight 187.24
Accessories

GHS Signal Word: Danger

SDS & Safety Documents