missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-1-naphthylmethylamine 98.0+%, TCI America™
Supplier: TCI America M09835G
Specifications
N-Methyl-1-naphthylmethylamine | |
Yellow | |
C12H13N | |
5 g | |
MQRIUFVBEVFILS-UHFFFAOYSA-N | |
N-methyl-1-naphthalen-1-ylmethanamine | |
84474 | |
≥98.0% (T) |
14489-75-9 | |
120°C | |
MFCD00144934 | |
1-(Methylaminomethyl)naphthalene | |
CNCC1=CC=CC2=CC=CC=C21 | |
171.243 | |
171.24 | |
Liquid |
Chemical Identifiers
14489-75-9 | |
171.243 | |
MQRIUFVBEVFILS-UHFFFAOYSA-N | |
84474 | |
CNCC1=CC=CC2=CC=CC=C21 |
C12H13N | |
MFCD00144934 | |
1-(Methylaminomethyl)naphthalene | |
N-methyl-1-naphthalen-1-ylmethanamine |
Safety and Handling
TSCA : No