missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-(1-methylpiperid-3-yl)methylamine, 90%, Maybridge

Chemical Identifiers

CAS 639078-61-8
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C8H18N2
Formula Weight 142.24
MDL Number MFCD05864531
Chemical Name or Material N-methyl-(1-methylpiperid-3-yl)methylamine
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC66146CB SDS Fisher Scientific Company
CC66146CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC66146DA SDS Fisher Scientific Company
CC66146DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

639078-61-8
100.0
C8H18N2
MFCD05864531
90%
89.5
90%
142.24
N-methyl-(1-methylpiperid-3-yl)methylamine

Specifications

142.24
100.0
MFCD05864531
Amber Glass Bottle
C8H18N2
90%
250mg
90%
89.5
639078-61-8
N-methyl-(1-methylpiperid-3-yl)methylamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger