Learn More
n-methyl-(1-methyl-1h-indol-7-yl)methylamine, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC66946CB
Specifications
N-methyl-(1-methyl-1H-indol-7-yl)methylamine | |
89.5 | |
90% | |
C11H14N2 | |
250mg | |
JERMRTDADLXHSD-UHFFFAOYSA-N | |
N-methyl-1-(1-methylindol-7-yl)methanamine | |
33589520 | |
90% |
709649-75-2 | |
100.0 | |
Amber Glass Bottle | |
MFCD11506353 | |
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine | |
CNCC1=CC=CC2=C1N(C=C2)C | |
174.247 | |
174.25 |
Chemical Identifiers
709649-75-2 | |
174.247 | |
JERMRTDADLXHSD-UHFFFAOYSA-N | |
33589520 | |
CNCC1=CC=CC2=C1N(C=C2)C |
C11H14N2 | |
MFCD11506353 | |
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine | |
N-methyl-1-(1-methylindol-7-yl)methanamine |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger