Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-1-(2-thienyl)ethylamine hydrochloride

$263.99 - $263.99

Chemical Identifiers

Chemical Name or Material N-Methyl-1-(2-thienyl)ethylamine hydrochloride
CAS 174316-28-0
Molecular Formula C7H11NS·HCl
Formula Weight 177.6g/mol
MDL Number MFCD03306017
Synonym alpha,N-Dimethyl-2-thiophenemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51109MD

 
alfa aesar™
H51109-MD
Each for $263.99
Description & Specifications

Specifications

Quantity 250mg
Chemical Name or Material N-Methyl-1-(2-thienyl)ethylamine hydrochloride
CAS 174316-28-0
Molecular Formula C7H11NS·HCl
Formula Weight 177.6g/mol
MDL Number MFCD03306017
Synonym alpha,N-Dimethyl-2-thiophenemethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.